Learn More
Medchemexpress LLC RVG-Cys acetate | 99.7% | 3369.77 (free acid) | 10 MG
SDP
Catalog No. 5000398581
Encompass_Preferred
RVG-Cys (RVG29-Cys) acetate is a peptide derived from rabies virus glycoprotein (RVG29) with Cys attached to facilitate subsequent conjugation. It enhances the specific targeted delivery of proteins in brain tissue and neurons.
  • Peptide derived from rabies virus glycoprotein (RVG29)
  • Includes Cys for subsequent conjugation
  • Enhances specific targeted delivery of proteins
  • Delivers to brain tissue and neurons
Catalog No. 50-003-98581 Supplier Medchemexpress LLC Supplier No. HYP5623A110MG
RVG-Cys (RVG29-Cys) acetate is a peptide derived from rabies virus glycoprotein (RVG29) with Cys attached to facilitate subsequent conjugation. It enhances the specific targeted delivery of proteins in brain tissue and neurons.
  • Peptide derived from rabies virus glycoprotein (RVG29)
  • Includes Cys for subsequent conjugation
  • Enhances specific targeted delivery of proteins
  • Delivers to brain tissue and neurons