Learn More
Medchemexpress LLC RVG-Cys acetate | 99.7% | 3369.77 (free acid) | 5 MG
SDP
Catalog No. 5000397000
Encompass_Preferred
RVG-Cys acetate is a peptide derived from rabies virus glycoprotein (RVG29). It is designed with a Cys attachment to facilitate subsequent conjugation and enhances the specific targeted delivery of proteins in brain tissue and neurons. This product is for research use only.
  • Peptide derived from rabies virus glycoprotein (RVG29)
  • Cys attached for subsequent conjugation
  • Enhances specific targeted delivery of proteins
  • Targets brain tissue and neurons
  • For research use only
Catalog No. 50-003-97000 Supplier Medchemexpress LLC Supplier No. HYP5623A15MG
RVG-Cys acetate is a peptide derived from rabies virus glycoprotein (RVG29). It is designed with a Cys attachment to facilitate subsequent conjugation and enhances the specific targeted delivery of proteins in brain tissue and neurons. This product is for research use only.
  • Peptide derived from rabies virus glycoprotein (RVG29)
  • Cys attached for subsequent conjugation
  • Enhances specific targeted delivery of proteins
  • Targets brain tissue and neurons
  • For research use only