Learn More
Cayman Chemical RVG PeptIDtrIfluoROCTAte 10mg
SDP
Catalog No. 502945547
Encompass_Preferred
A peptide nAChR antagonist (IC50 = 2.5 µM); LNPs conjugated to RVG peptide selectively traffic to the brain over the lungs and liver in rats; LNPs conjugated to RVG peptide and encapsulating DFO reduce iron levels in the striatum and substantia nigra, as well as decrease levels of ROS and MDA and increase levels of SOD in the substantia nigra in a mouse model of MPTP-induced Parkinson's disease; RVG-containing and DFO-encapsulating LNPs increase the latency to fall in the rotarod test in the same model of MPTP-induced Parkinson's disease
Catalog No. 50-294-5547 Supplier Cayman Chemical Supplier No. 4160610 MG
A peptide nAChR antagonist (IC50 = 2.5 µM); LNPs conjugated to RVG peptide selectively traffic to the brain over the lungs and liver in rats; LNPs conjugated to RVG peptide and encapsulating DFO reduce iron levels in the striatum and substantia nigra, as well as decrease levels of ROS and MDA and increase levels of SOD in the substantia nigra in a mouse model of MPTP-induced Parkinson's disease; RVG-containing and DFO-encapsulating LNPs increase the latency to fall in the rotarod test in the same model of MPTP-induced Parkinson's disease