Learn More
Cygnus Technologies RVLP Stock Solution, Non-Infectious, BSL-1 Compatible, 2.0 mL
Catalog No. NC2236253
Encompass
The RVLP Stock Solution is a key component of the MockV RVLP Kit, designed for efficient quantification of Retrovirus-Like Particle removal during biopharmaceutical manufacturing. This non-infectious, CHO-cell derived solution allows for safe viral clearance studies in BSL-1 labs, offering a cost-effective alternative to conventional viral clearance methods, thus optimizing process development and ensuring product safety.
This item is not returnable. View return policy
Catalog No. NC2236253 Supplier Cygnus Technologies Supplier No. M2311
The RVLP Stock Solution is a key component of the MockV RVLP Kit, designed for efficient quantification of Retrovirus-Like Particle removal during biopharmaceutical manufacturing. This non-infectious, CHO-cell derived solution allows for safe viral clearance studies in BSL-1 labs, offering a cost-effective alternative to conventional viral clearance methods, thus optimizing process development and ensuring product safety.