Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbix Biosystems RVP4 (H3) Control Panel

Catalog No. 21377306 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
Sample Type:
Nasal Swab
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
21-377-306 Nasal Swab
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-377-306 Supplier Microbix Biosystems Supplier No. CPKS00008
Add to Cart
Add to Cart

Intended for use with the Aptitude Metrix COVID/Flu test, formulated with inactivated SARS-CoV-2 whole genome nucleic acid and inactivated Influenza A, Influenza B, and Respiratory Syncytial Virus.

  • Optimal for Point of Care (POC) near-patient testing workflows.
  • Stable for up to 2 years when stored at 2-30°C.
  • Desiccated on a Copan FLOQSwab™.
  • Can be eluted in your choice of media.
  • Suitable for storage and distribution under standard supply chain conditions.
  • Demonstrates consistent performance across production lots.
  • Includes desiccated positive and negative control swabs designed to monitor NAAT performance for respiratory viral detection workflows.
TRUSTED_SUSTAINABILITY

Specifications

Sample Type Nasal Swab
Type Control Panel
Disposable Single-use
Quantity 2
Stability 2 to 30 °C
Sterility Sterile
Form Swab
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.