Learn More
NEOGEN Rappaport-Vassiliadis Medium with Soya (RVS) (ISO), 5 kg
SDP
Catalog No. 502015544
Encompass_Preferred
Rappaport-Vassiliadis Medium with Soya is used as a selective enrichment medium for the detection of Salmonella spp. in food, animal feed and in environmental samples from the food production area as described in ISO 6579-1:2017. RVS is not intended for use in the diagnosis of disease or other conditions in humans.Rappaport et al. originally described the broth, which was later modified by Vassiliadis et al. and optimized by van Schothorst et al. Enzymatic digest of soya provides a source of nitrogen and amino acids and sodium chloride provides osmotic pressure. The potassium phosphate components act as a buffer. Magnesium chloride and malachite green act as selective agents against non-target microorganisms. The formulation is very hygroscopic and will produce a slight exothermic reaction when mixed with water.For more information visit https://www.neogen.com/solutions/Microbiology/rappaport-vassiliadis-medium-with-soya
Catalog No. 50-201-5544 Supplier NEOGEN Supplier No. 700004528
Rappaport-Vassiliadis Medium with Soya is used as a selective enrichment medium for the detection of Salmonella spp. in food, animal feed and in environmental samples from the food production area as described in ISO 6579-1:2017. RVS is not intended for use in the diagnosis of disease or other conditions in humans.Rappaport et al. originally described the broth, which was later modified by Vassiliadis et al. and optimized by van Schothorst et al. Enzymatic digest of soya provides a source of nitrogen and amino acids and sodium chloride provides osmotic pressure. The potassium phosphate components act as a buffer. Magnesium chloride and malachite green act as selective agents against non-target microorganisms. The formulation is very hygroscopic and will produce a slight exothermic reaction when mixed with water.For more information visit https://www.neogen.com/solutions/Microbiology/rappaport-vassiliadis-medium-with-soya