Learn More
Apexbio Technology LLC RVX-208 1044870-39-4 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B14995.1
Encompass_Preferred
RVX-208 (CAS 1044870-39-4) is a small-molecule inhibitor targeting BET (bromodomain and extra-terminal) proteins It is designed to selectively inhibit the binding of BET bromodomains to acetylated lysine residues thereby modulating transcriptional regulation RVX-208 exerts its biological activity primarily through competitive inhibition of the second bromodomain (BD2) of BET proteins In in vitro studies using human hepatic (HepG2) cells RVX-208 demonstrates BD2 inhibitory activity with an IC50 value of 0 51 M and lower affinity for BD1 (IC50 87 M) Based on these pharmacological properties RVX-208 holds research potential in lipid metabolism cholesterol efflux and atherosclerotic disease processes
Catalog No. 50-000-92045