Learn More
Medchemexpress LLC HY-16652 50mg , RVX-208 Apabetalone;RVX000222 CAS:1044870-39-4 Purity:98%
SDP
Catalog No. 501493752
Encompass_Preferred
Medchemexpress, HY-16652 50mg RVX-208 Apabetalone;RVX000222 CAS:1044870-39-4 Formula:C20H22N2O5 IC50: 510±41 nM (BD2), 87±10 μM (BD1) Purity:98% RVX-208 is an inhibitor of BET transcriptional regulators with selectivity for the second bromodomain. The IC 50 s are 87±10 μM and 0.51±0.041 μM for BD1 and BD2 , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3752 Supplier Medchemexpress LLC Supplier No. HY1665250MG
Medchemexpress, HY-16652 50mg RVX-208 Apabetalone;RVX000222 CAS:1044870-39-4 Formula:C20H22N2O5 IC50: 510±41 nM (BD2), 87±10 μM (BD1) Purity:98% RVX-208 is an inhibitor of BET transcriptional regulators with selectivity for the second bromodomain. The IC 50 s are 87±10 μM and 0.51±0.041 μM for BD1 and BD2 , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.