Learn More
Medchemexpress LLC RVX-297 100mg | 1044871-04-6 | 100 MG
SDP
Catalog No. 5000228960
Encompass_Preferred
RVX-297 is a BD2-selective BET bromodomain inhibitor provided as a research compound for studying bromodomain-mediated regulation of gene expression and inflammation. It is supplied as a powder with defined purity and storage recommendations for laboratory use.
  • BD2-selective BET bromodomain inhibitor for in vitro and in vivo research.
  • Shown to suppress inflammatory gene expression in multiple immune cell models.
  • Supplied as a powder with specified storage conditions to maintain stability.
  • Common applications include studies of inflammation, autoimmunity, and epigenetic regulation.
Catalog No. 50-002-28960 Supplier Medchemexpress LLC Supplier No. HY114504100MG
RVX-297 is a BD2-selective BET bromodomain inhibitor provided as a research compound for studying bromodomain-mediated regulation of gene expression and inflammation. It is supplied as a powder with defined purity and storage recommendations for laboratory use.
  • BD2-selective BET bromodomain inhibitor for in vitro and in vivo research.
  • Shown to suppress inflammatory gene expression in multiple immune cell models.
  • Supplied as a powder with specified storage conditions to maintain stability.
  • Common applications include studies of inflammation, autoimmunity, and epigenetic regulation.