Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RVX-297 10mg | 1044871-04-6 | 10 MG
SDP

Catalog No. 5000229326
Encompass_Preferred

RVX-297 is a small-molecule BD2-selective BET bromodomain inhibitor used in preclinical research to study epigenetic regulation of inflammation and autoimmune responses. It is supplied as a powder for laboratory research use only.

  • BD2-selective BET bromodomain inhibitor.
  • Reported IC50s: BRD2(BD2) 0.08 μM; BRD3(BD2) 0.05 μM; BRD4(BD2) 0.02 μM.
  • CAS number 1044871-04-6.
  • Molecular formula C24H29N3O4; molecular weight 423.50 g/mol.
  • Typical reported purity ~95%.
  • Powder storage: -20°C for long-term stability.
  • For research use only; not for human use.

Catalog No. 50-002-29326 Supplier Medchemexpress LLC Supplier No. HY11450410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RVX-297 is a small-molecule BD2-selective BET bromodomain inhibitor used in preclinical research to study epigenetic regulation of inflammation and autoimmune responses. It is supplied as a powder for laboratory research use only.

  • BD2-selective BET bromodomain inhibitor.
  • Reported IC50s: BRD2(BD2) 0.08 μM; BRD3(BD2) 0.05 μM; BRD4(BD2) 0.02 μM.
  • CAS number 1044871-04-6.
  • Molecular formula C24H29N3O4; molecular weight 423.50 g/mol.
  • Typical reported purity ~95%.
  • Powder storage: -20°C for long-term stability.
  • For research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.