Learn More
Medchemexpress LLC RVX-297 25mg | 1044871-04-6 | 25 MG
SDP
Catalog No. 5000230001
Encompass_Preferred
RVX-297 is a BD2-selective BET bromodomain inhibitor supplied for laboratory research. It is a small-molecule compound characterized for activity against BET family bromodomains and is used in preclinical studies of inflammation and autoimmune disease.
  • Selective BD2 bromodomain inhibition for research applications.
  • Chemical formula C24H29N3O4 and molecular weight 423.50 g/mol.
  • Supplied as a dry powder with ≥95% purity.
  • Available in small-scale vials suitable for laboratory experiments.
  • Recommended cold storage to preserve stability (powder and solution guidelines).
Catalog No. 50-002-30001 Supplier Medchemexpress LLC Supplier No. HY11450425MG
RVX-297 is a BD2-selective BET bromodomain inhibitor supplied for laboratory research. It is a small-molecule compound characterized for activity against BET family bromodomains and is used in preclinical studies of inflammation and autoimmune disease.
  • Selective BD2 bromodomain inhibition for research applications.
  • Chemical formula C24H29N3O4 and molecular weight 423.50 g/mol.
  • Supplied as a dry powder with ≥95% purity.
  • Available in small-scale vials suitable for laboratory experiments.
  • Recommended cold storage to preserve stability (powder and solution guidelines).