Learn More
Medchemexpress LLC RVX-297 50mg | 1044871-04-6 | 423.50 | C24H29N3O4 | 50 MG
SDP
Catalog No. 5000308018
Encompass_Preferred
RVX-297 is a potent, orally active BD2-selective BET bromodomain inhibitor used in preclinical research to study inflammation and autoimmune disease mechanisms. It preferentially binds BD2 domains of BET proteins and suppresses inflammatory gene expression.
  • BD2-selective BET bromodomain inhibitor for mechanistic studies.
  • Orally active in preclinical models of inflammation and autoimmunity.
  • Potent activity: BRD2(BD2) IC50 = 0.08 μM, BRD3(BD2) = 0.05 μM, BRD4(BD2) = 0.02 μM.
  • High purity (95.0%) suitable for in vitro and in vivo research.
  • Stable solid: powder storage recommended at low temperatures for long-term stability.
Catalog No. 50-003-08018 Supplier Medchemexpress LLC Supplier No. HY11450450MG
RVX-297 is a potent, orally active BD2-selective BET bromodomain inhibitor used in preclinical research to study inflammation and autoimmune disease mechanisms. It preferentially binds BD2 domains of BET proteins and suppresses inflammatory gene expression.
  • BD2-selective BET bromodomain inhibitor for mechanistic studies.
  • Orally active in preclinical models of inflammation and autoimmunity.
  • Potent activity: BRD2(BD2) IC50 = 0.08 μM, BRD3(BD2) = 0.05 μM, BRD4(BD2) = 0.02 μM.
  • High purity (95.0%) suitable for in vitro and in vivo research.
  • Stable solid: powder storage recommended at low temperatures for long-term stability.