Learn More
Pyramex™ RVZ24 Series - Safety Vest
Catalog No. 19555559 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Hi-vis Lime
Hi-vis Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-555-559 Hi-vis Lime Small
19-555-558 Hi-vis Lime Medium
19-555-557 Hi-vis Lime Large
19-555-571 Hi-vis Lime X-Large
19-555-567 Hi-vis Lime 2X-Large
19-555-568 Hi-vis Lime 3X-Large
19-555-569 Hi-vis Lime 4X-Large
19-555-570 Hi-vis Lime 5X-Large
19-555-574 Hi-vis Orange Small
19-555-573 Hi-vis Orange Medium
19-555-572 Hi-vis Orange Large
19-555-584 Hi-vis Orange X-Large
19-555-582 Hi-vis Orange 2X-Large
19-555-583 Hi-vis Orange 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Catalog No. 19-555-559 Supplier Pyramex™ Supplier No. RVZ2410S
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pyramex™ RVZ24 Series - Safety Vest with ANSI/ISEA 107-2015 safety compliance
  • Hi-Vis Surveyor's Vest
  • Front, Zipper Closure
  • Lightweight polyester solid front, and mesh back
  • One 4 in. contrasting material stripe applied horizontally with 2 in. silver reflective stripe centered inside contrasting stripe
  • Two, 4 in. contrasting material stripes with 2 in. silver reflective stripe centered inside contrasting stripe run vertically from bottom stripe, over shoulders to the bottom of the vest
  • Chest Pockets: Outer left of vest divided into four sections sewn on top of full size pocket (5.5 x 5.5 in.). Outer right of vest gusseted with flap closure and hook & loop tab (4 x 5.5 in.)
  • Outside Pockets: One on both the left, and right sides with flap closure and hook & loop tab (7.5 x 7.5 in.)
  • Inside Pockets: One on both the left, and the right sides (7.5 x 7.5 in.)
  • Flashlight Pocket: Inside right (2 x 7.5 in.)
  • Double tacked badge/mic holder located 3 in. from the center shoulder seam on both sides (1 x 4 in.)
  • Individually packaged
  • 50 vests per case.
Specifications
Closure Type Zipper
Certifications/Compliance ANSI/ISEA 107-2015, Type R - Class 2
Color Hi-vis Lime
Size Small
Material Polyester
Size (English) Chest 34-36 in.
No. of Pockets 8
Tape Type China Brand Silver