Learn More
Cayman Chemical RW2 (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502916111
Encompass_Preferred
An antimicrobial peptide; active against E. coli and S. aureus (IC50s = 730 and 210 µM, respectively); induces hemolysis of isolated ovine erythrocytes
Catalog No. 50-291-6111 Supplier Cayman Chemical Supplier No. 3749210 MG
An antimicrobial peptide; active against E. coli and S. aureus (IC50s = 730 and 210 µM, respectively); induces hemolysis of isolated ovine erythrocytes