Learn More
Cayman Chemical RW3trIfluoroacetate salt 50mg
SDP
Catalog No. 502916113
Encompass_Preferred
An antimicrobial peptide; active against E. coli and S. aureus (IC50s = 16 and 8 µM, respectively); induces hemolysis of isolated ovine erythrocytes
Catalog No. 50-291-6113 Supplier Cayman Chemical Supplier No. 3749350 MG
An antimicrobial peptide; active against E. coli and S. aureus (IC50s = 16 and 8 µM, respectively); induces hemolysis of isolated ovine erythrocytes