Learn More
Cayman Chemical RW4trIfluoroacetate salt 10mg
SDP
Catalog No. 502916115
Encompass_Preferred
An antimicrobial peptide; active against E. coli and S. aureus (IC50s = 9.6 and 5.1 µM, respectively); induces hemolysis of isolated ovine erythrocytes
Catalog No. 50-291-6115 Supplier Cayman Chemical Supplier No. 3749410 MG
An antimicrobial peptide; active against E. coli and S. aureus (IC50s = 9.6 and 5.1 µM, respectively); induces hemolysis of isolated ovine erythrocytes