Learn More
Nanoentek Inc ADAM rWBC Kit
SDP
Catalog No. NC1316634
Encompass
ADAM rWBC series are based on the staining methods for the DNA of mammalian cells using the fluorescent dye, Propidium Iodide (PI). When used with RNase, PI stains only cellular DNA as the nucleic acid dye. In the case of blood samples, white blood cells (with DNA-containing nuclei) are stained while platelets and red blood cells, (without nuclei) are not stained. 
Catalog No. NC1316634 Supplier Nanoentek Inc Supplier No. AD1K050
ADAM rWBC series are based on the staining methods for the DNA of mammalian cells using the fluorescent dye, Propidium Iodide (PI). When used with RNase, PI stains only cellular DNA as the nucleic acid dye. In the case of blood samples, white blood cells (with DNA-containing nuclei) are stained while platelets and red blood cells, (without nuclei) are not stained. 