Learn More
Aviva Systems Biology Corp RWDD1 Antibody - middle region (ARP59212_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59212_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RWDD1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RWDD1. Alternate names: CGI-24, PTD013ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-618