Learn More
Medchemexpress LLC HY-110086 5mg Medchemexpress, RWJ 50271 CAS:162112-37-0 Purity:>98%
SDP
Catalog No. 501932345
Encompass_Preferred
Medchemexpress, HY-110086 5mg RWJ 50271 CAS:162112-37-0 RWJ 50271 is an selective inhibitor of lymphocyte function-associated antigen-1/intercellular adhesion molecule-1(LFA-1/ICAM-1) interaction with an IC 50 of 5.0 μM (HL60 cells) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2345 Supplier Medchemexpress LLC Supplier No. HY1100865MG
Medchemexpress, HY-110086 5mg RWJ 50271 CAS:162112-37-0 RWJ 50271 is an selective inhibitor of lymphocyte function-associated antigen-1/intercellular adhesion molecule-1(LFA-1/ICAM-1) interaction with an IC 50 of 5.0 μM (HL60 cells) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.