Learn More
Medchemexpress LLC RWJ-56110 dihydrochloride | 2387505-58-8 | 99.9% | 863.65 | 1 ML
SDP
Catalog No. 5000346962
Encompass_Preferred
RWJ-56110 dihydrochloride is a potent, selective, peptide-mimetic inhibitor that targets PAR-1 activation and internalization, with a binding IC50 of 0.44 μM. It does not affect PAR-2, PAR-3, or PAR-4. This compound inhibits the aggregation of human platelets induced by SFLLRN-NH2 (IC50=0.16 μM) and thrombin (IC50=0.34 μM), demonstrating selectivity compared to U46619. Additionally, RWJ-56110 dihydrochloride blocks angiogenesis by preventing new vessel formation in vivo and has been shown to induce cell apoptosis.
  • It is a potent, selective, peptide-mimetic inhibitor of PAR-1 activation and internalization.
  • It shows no effect on PAR-2, PAR-3, or PAR-4.
  • It inhibits the aggregation of human platelets induced by SFLLRN-NH2 and thrombin.
  • It blocks angiogenesis and the formation of new vessels in vivo.
  • It induces cell apoptosis.
Catalog No. 50-003-46962 Supplier Medchemexpress LLC Supplier No. HY108556A1MG
RWJ-56110 dihydrochloride is a potent, selective, peptide-mimetic inhibitor that targets PAR-1 activation and internalization, with a binding IC50 of 0.44 μM. It does not affect PAR-2, PAR-3, or PAR-4. This compound inhibits the aggregation of human platelets induced by SFLLRN-NH2 (IC50=0.16 μM) and thrombin (IC50=0.34 μM), demonstrating selectivity compared to U46619. Additionally, RWJ-56110 dihydrochloride blocks angiogenesis by preventing new vessel formation in vivo and has been shown to induce cell apoptosis.
  • It is a potent, selective, peptide-mimetic inhibitor of PAR-1 activation and internalization.
  • It shows no effect on PAR-2, PAR-3, or PAR-4.
  • It inhibits the aggregation of human platelets induced by SFLLRN-NH2 and thrombin.
  • It blocks angiogenesis and the formation of new vessels in vivo.
  • It induces cell apoptosis.