Learn More
Abcam RWJ-60475 (AM)3, CD45 tyrosine phosphatase inhibitor, 1MG-SLD
Catalog No. 502706463
Encompass_Preferred
MW 620.4 g/mol, Purity >95%. CD45 tyrosine phosphatase inhibitor (IC₅₀ = 2 μM). Cell permeable. Inhibits ADP induced platelet aggregation and IL-2 production in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6463 Supplier Abcam Supplier No. AB1417291MGSLD
MW 620.4 g/mol, Purity >95%. CD45 tyrosine phosphatase inhibitor (IC₅₀ = 2 μM). Cell permeable. Inhibits ADP induced platelet aggregation and IL-2 production in vitro.
The product is subject to the following: Abcam Restricted Use Statement