Learn More
Medchemexpress LLC HY-15505 5mg Medchemexpress, RWJ-67657 CAS:215303-72-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY155055MG
Encompass_Preferred
Medchemexpress, HY-15505 5mg RWJ-67657 CAS:215303-72-3 RWJ-67657 (JNJ 3026582) is an orally active and selective p38α and p38β MAPK inhibitor with IC50s of 1 and 11 μM, respectively. RWJ-67657 displays no activity at p38γ and p38δ, and exhibits cardio protective effect. Anti-inflammatory and anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9945