Learn More
Medchemexpress LLC Rx 801077 hydrochloride | 89196-95-2 | 99.8% | 222.67 g/mol | C11H11ClN2O | 50 MG
SDP
Catalog No. 5000212252
Encompass_Preferred
RX 801077 hydrochloride (2 BFI) is a selective imidazoline I2 receptor agonist used in neuroscience research. It exhibits nanomolar binding affinity to I2 receptors and has been reported to show anti-inflammatory and neuroprotective effects relevant to traumatic brain injury and receptor pharmacology studies.
  • Selective imidazoline I2 receptor agonist.
  • High purity: 99.8%.
  • Hydrochloride salt for improved stability and handling.
  • Suitable for in vitro and in vivo pharmacology research.
  • Molecular weight: 222.67 g/mol.
  • Reported anti-inflammatory and neuroprotective activity.
  • Available as a 50 MG research-size package.
Catalog No. 50-002-12252 Supplier Medchemexpress LLC Supplier No. HY10090450MG
RX 801077 hydrochloride (2 BFI) is a selective imidazoline I2 receptor agonist used in neuroscience research. It exhibits nanomolar binding affinity to I2 receptors and has been reported to show anti-inflammatory and neuroprotective effects relevant to traumatic brain injury and receptor pharmacology studies.
  • Selective imidazoline I2 receptor agonist.
  • High purity: 99.8%.
  • Hydrochloride salt for improved stability and handling.
  • Suitable for in vitro and in vivo pharmacology research.
  • Molecular weight: 222.67 g/mol.
  • Reported anti-inflammatory and neuroprotective activity.
  • Available as a 50 MG research-size package.