Learn More
Biotang Inc Autoradiography Film, 8X10in 100pk, Blue Bio Film, For blotting and sequencing; optimized for chemilluminescence, Compatible with automatic or manual processors,Extremely Low Backgrounds (Blue) Publishable Grade Autoradiographs, Made in USA
Catalog No. 507514993 Shop All BIOTANG Inc Products
Encompass_Preferred
Autoradiography Film, 8X10in 100pk, Blue Bio Film, For blotting and sequencing; optimized for chemilluminescence, Compatible with automatic or manual processors,Extremely Low Backgrounds (Blue) Publishable Grade Autoradiographs, Made in USA
Catalog No. 50-751-4993 Supplier Biotang Inc Supplier No. AFJ08F
Autoradiography Film, 8X10in 100pk, Blue Bio Film, For blotting and sequencing; optimized for chemilluminescence, Compatible with automatic or manual processors,Extremely Low Backgrounds (Blue) Publishable Grade Autoradiographs, Made in USA