Learn More
Med Vet International Gebauer'S Ethyl Chloride Spray, Medium, 3.5oz
Catalog No. 502827948
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Gebauer's Ethyl Chloride is a vapocoolant (skin refrigerant) intended for topical application to control pain associated with injections, starting IVs and venipuncture, minor surgical procedures (such as lancing boils or incision and drainage of small abscesses), and the temporary relief of minor sports injuries.
Catalog No. 50-282-7948 Supplier Med Vet International Supplier No. RXETHYLCHLO
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Gebauer's Ethyl Chloride is a vapocoolant (skin refrigerant) intended for topical application to control pain associated with injections, starting IVs and venipuncture, minor surgical procedures (such as lancing boils or incision and drainage of small abscesses), and the temporary relief of minor sports injuries.