Learn More
Med Vet International Panakare Powder, Digestive Aid, Pancreatic Enzyme Concentrate, 8oz
Catalog No. 502827933
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
PanaKare Plus is for use as a digestive aid in replacement therapy in dogs and cats where digestion of proteins, carbohydrates, and fats is inadequate due to exocrine pancreatic insufficiency(EPI). EPI is also known as Maldigestion. This is a condition that occurs when the cells of the pancreas that produce digestive enzymes become damaged or lacking. This results in malnutrition and can occur in any breed or age. Common symptoms include: Progressive weight loss Semi-formed stools intermittent vomiting Coprophagia PanaKare Plus is a pancreatic enzyme concentrate of porcine origin. For animal use only. 8oz bottle.
Catalog No. 50-282-7933 Supplier Med Vet International Supplier No. RXPANAKARE8
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
PanaKare Plus is for use as a digestive aid in replacement therapy in dogs and cats where digestion of proteins, carbohydrates, and fats is inadequate due to exocrine pancreatic insufficiency(EPI). EPI is also known as Maldigestion. This is a condition that occurs when the cells of the pancreas that produce digestive enzymes become damaged or lacking. This results in malnutrition and can occur in any breed or age. Common symptoms include: Progressive weight loss Semi-formed stools intermittent vomiting Coprophagia PanaKare Plus is a pancreatic enzyme concentrate of porcine origin. For animal use only. 8oz bottle.