Learn More
Medchemexpress LLC (RXR)4XB TFA | 96.6% | 1904.37 | 1 MG
SDP
Catalog No. 5000422870
Encompass_Preferred
(RXR)4XB TFA is a cell-penetrating peptide that binds phosphorodiamidate morpholino oligomers (PMOs), forming peptide-conjugated PMOs (PPMOs), and enhances the delivery of PMO into bacterial cells. The (RXR)4XB TFA-PMO conjugate is effective in preventing biofilm formation, inhibiting *Pseudomonas aeruginosa* with an MIC50 of 0.5 to 16 μM, and reducing bacterial burden in mouse acute pneumonia models.
  • Forms peptide-conjugated PMOs (PPMOs)
  • Enhances delivery of PMO into bacterial cells
  • Prevents biofilm formation
  • Inhibits *Pseudomonas aeruginosa*
  • Reduces bacterial burden in mouse acute pneumonia models
Catalog No. 50-004-22870 Supplier Medchemexpress LLC Supplier No. HYP10559A1MG
(RXR)4XB TFA is a cell-penetrating peptide that binds phosphorodiamidate morpholino oligomers (PMOs), forming peptide-conjugated PMOs (PPMOs), and enhances the delivery of PMO into bacterial cells. The (RXR)4XB TFA-PMO conjugate is effective in preventing biofilm formation, inhibiting *Pseudomonas aeruginosa* with an MIC50 of 0.5 to 16 μM, and reducing bacterial burden in mouse acute pneumonia models.
  • Forms peptide-conjugated PMOs (PPMOs)
  • Enhances delivery of PMO into bacterial cells
  • Prevents biofilm formation
  • Inhibits *Pseudomonas aeruginosa*
  • Reduces bacterial burden in mouse acute pneumonia models