Learn More
Cayman Chemical RXR aRetInoId X RECPTR a 100uL
SDP
Catalog No. 502948545
Encompass_Preferred
An affinity probe for ganglioside GM3 binding partners
Catalog No. 50-294-8545 Supplier Cayman Chemical Supplier No. 32825100 UL
An affinity probe for ganglioside GM3 binding partners