Learn More
Enzo Life Sciences Retinoid X receptor beta (human), (recombinant) (50 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE1270050
Encompass_Preferred
Alternative name: RXR beta. Concentration: 2 mg/ml total protein (~0.6 mg/ml RXRß). Formulation: Liquid. In 20 mM HEPES, pH 7.9, containing 0.1 M KCl, 0.2 mM EDTA, 0.5 mM DTT and 20% glycerol. Source: Produced in Sf9 insect cells. Full length RXRß (rH2RIIBP). Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-1938