Learn More
Cayman Chemical RXR RetInoId X receptr100uL
SDP
Catalog No. 502946585
Encompass_Preferred
A KCC2 cotransporter activator (EC50 616 nM for chloride transport in NG108-cl cells) selective for KCC2 over GABAA receptors at 50 UM increases the rate of chloride accumulation in spinal slices isolated from a rat model of PNI and BDNF control slices increases mechanical withdrawal thresholds in a rat model of PNI and reverses morphine-induced hyperalgesia in rats at 100 mg/kg
Catalog No. 50-294-6585 Supplier Cayman Chemical Supplier No. 32826100 UL
A KCC2 cotransporter activator (EC50 616 nM for chloride transport in NG108-cl cells) selective for KCC2 over GABAA receptors at 50 UM increases the rate of chloride accumulation in spinal slices isolated from a rat model of PNI and BDNF control slices increases mechanical withdrawal thresholds in a rat model of PNI and reverses morphine-induced hyperalgesia in rats at 100 mg/kg