Learn More
Aviva Systems Biology Corp RXRA Antibody - N-terminal region (ARP34060_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34060_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RXRA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RXRA. Alternate names: FLJ16020, FLJ16733, MGC102720, NR2B1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-55234