Learn More
Aviva Systems Biology Corp RXRB Antibody - N-terminal region (ARP45638_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45638_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RXRB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RXRB. Alternate names: DAUDI6, H-2RIIBP, MGC1831, NR2B2, RCoR-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59088