Learn More
Med Vet International Hyalovet 10/mL, 2mL Vial
Catalog No. 502827878
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Hyaluronic sodium injection indicated for the intra-articular treatment of carpal or fetlock joint dysfunction in horses due to acute or chronic, non-infectious synovitis associated with equine osteoarthritis. Each filled 2 mL glass vial contains: - Hyaluronate Sodium, 20 mg - Sodium Chloride, 17 mg - Monobasic Sodium Phosphate, 0.1 mg - Dibasic Sodium Phosphate, 1.2 mg - Water for Injection, q.s., 2 mL
Catalog No. 50-282-7878 Supplier Med Vet International Supplier No. RXHYALOV2VI
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Hyaluronic sodium injection indicated for the intra-articular treatment of carpal or fetlock joint dysfunction in horses due to acute or chronic, non-infectious synovitis associated with equine osteoarthritis. Each filled 2 mL glass vial contains: - Hyaluronate Sodium, 20 mg - Sodium Chloride, 17 mg - Monobasic Sodium Phosphate, 0.1 mg - Dibasic Sodium Phosphate, 1.2 mg - Water for Injection, q.s., 2 mL