Learn More
Med Vet International Merck Mometamax, 30gm, 6/Box
Catalog No. 502828263
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
MOMETAMAX is a combination of gentamicin, mometasone, and clotrimazole. Used to treat inflammation and bacterial infections of the ear. Gentamicin is a broad-spectrum antibiotic, mometasone is a potent steroid with minimal systemic absorption, and clotrimazole has antifungal properties. Dropper bottle includes longer tip for easier administration.
Catalog No. 50-282-8263 Supplier Med Vet International Supplier No. RXMOMETAMAX30
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
MOMETAMAX is a combination of gentamicin, mometasone, and clotrimazole. Used to treat inflammation and bacterial infections of the ear. Gentamicin is a broad-spectrum antibiotic, mometasone is a potent steroid with minimal systemic absorption, and clotrimazole has antifungal properties. Dropper bottle includes longer tip for easier administration.