Learn More
Med Vet International Otomax Ointment, 15g Tube
Catalog No. 502828000
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Combination steroid, antibacterial, and antifungal ointment used to treat acute and chronic otitis. Active ingredients are Betamethasone Valerate, Gentamicin Sulfate and Clotrimazole. For otic use in dogs only.
Catalog No. 50-282-8000 Supplier Med Vet International Supplier No. RXOTOMAX15TUBE
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Combination steroid, antibacterial, and antifungal ointment used to treat acute and chronic otitis. Active ingredients are Betamethasone Valerate, Gentamicin Sulfate and Clotrimazole. For otic use in dogs only.