Learn More
Med Vet International Otomax Ointment, 30gm
Catalog No. 502828479
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Otomax is indicated for the treatment of canine acute and chronic otitis externa associated with yeast (Malassezia pachydermatis, formerly Pityrosporum canis)and/or bacteria susceptible to gentamicin. Effective against otitis externa perpetuated by both yeast and bacteria. The consistency of a solution with the prolonged contact time of an ointment. Colorless ointment allows for accurate treatment evaluation.
Catalog No. 50-282-8479 Supplier Med Vet International Supplier No. RXOTOMAX30S
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Otomax is indicated for the treatment of canine acute and chronic otitis externa associated with yeast (Malassezia pachydermatis, formerly Pityrosporum canis)and/or bacteria susceptible to gentamicin. Effective against otitis externa perpetuated by both yeast and bacteria. The consistency of a solution with the prolonged contact time of an ointment. Colorless ointment allows for accurate treatment evaluation.