Learn More
Med Vet International Mometamax Otic Suspension, 215gm
Catalog No. 502828262
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
MOMETAMAX is a combination of gentamicin, mometasone, and clotrimazole. Used to treat inflammation and bacterial infections of the ear. Gentamicin is a broad-spectrum antibiotic, mometasone is a potent steroid with minimal systemic absorption, and clotrimazole has antifungal properties. Dropper bottle includes longer tip for easier administration.
Catalog No. 50-282-8262 Supplier Med Vet International Supplier No. RXMOMETAMAX215
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
MOMETAMAX is a combination of gentamicin, mometasone, and clotrimazole. Used to treat inflammation and bacterial infections of the ear. Gentamicin is a broad-spectrum antibiotic, mometasone is a potent steroid with minimal systemic absorption, and clotrimazole has antifungal properties. Dropper bottle includes longer tip for easier administration.