Learn More
Med Vet International Neo-Sol 50, 100gm
Catalog No. 502828464
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Neo-Sol 50 is indicated for the treatment and control of colibacillosis (bacterial enteritis) caused by Escherichia coli susceptible to neomycin sulfate in cattle (excluding veal calves), swine, sheep and goats. It is also indicated for the control of mortality associated with Escherichia coli organisms susceptible to neomycin sulfate in growing turkeys. See package insert for complete product details.
Catalog No. 50-282-8464 Supplier Med Vet International Supplier No. RXNEOSOL50100
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Neo-Sol 50 is indicated for the treatment and control of colibacillosis (bacterial enteritis) caused by Escherichia coli susceptible to neomycin sulfate in cattle (excluding veal calves), swine, sheep and goats. It is also indicated for the control of mortality associated with Escherichia coli organisms susceptible to neomycin sulfate in growing turkeys. See package insert for complete product details.