Learn More
Med Vet International Pancreplus Tablets, 100ct.
Catalog No. 502828861
Encompass_Preferred
A pancreatic enzyme concentrate of porcine origin fortified with vitamins A, D and E. It is used as a digestive aid in replacement therapy where digestion of protein, carbohydrate and fat is inadequate due to exocrine pancreatic insufficiency. For use in dogs and cats. Each tablet contains a minimum: Lipase, 9, 000 USP units Protease, 57, 000 USP units Amylase, 64, 000 USP units Vitamin A, 1, 000 IU Vitamin D3, 100 IU Vitamin E, 10 IU
Catalog No. 50-282-8861 Supplier Med Vet International Supplier No. RXPANCREPLUST
A pancreatic enzyme concentrate of porcine origin fortified with vitamins A, D and E. It is used as a digestive aid in replacement therapy where digestion of protein, carbohydrate and fat is inadequate due to exocrine pancreatic insufficiency. For use in dogs and cats. Each tablet contains a minimum: Lipase, 9, 000 USP units Protease, 57, 000 USP units Amylase, 64, 000 USP units Vitamin A, 1, 000 IU Vitamin D3, 100 IU Vitamin E, 10 IU