Learn More
Med Vet International Pancreplus Powder, 12oz.
Catalog No. 502827932
Encompass_Preferred
A pancreatic enzyme concentrate of porcine origin fortified with vitamins A, D and E. It is used as a digestive aid in replacement therapy where digestion of protein, carbohydrate and fat is inadequate due to exocrine pancreatic insufficiency. For use in dogs and cats. Each 2.8 g (1 teaspoon) contains a minimum: Lipase, 71,400 USP units Protease, 388,000 USP units Amylase, 460,000 USP units Vitamin A, 1,000 IU Vitamin D3, 100 IU Vitamin E, 10 IU
Catalog No. 50-282-7932 Supplier Med Vet International Supplier No. RXPANCREPLUS12
A pancreatic enzyme concentrate of porcine origin fortified with vitamins A, D and E. It is used as a digestive aid in replacement therapy where digestion of protein, carbohydrate and fat is inadequate due to exocrine pancreatic insufficiency. For use in dogs and cats. Each 2.8 g (1 teaspoon) contains a minimum: Lipase, 71,400 USP units Protease, 388,000 USP units Amylase, 460,000 USP units Vitamin A, 1,000 IU Vitamin D3, 100 IU Vitamin E, 10 IU