Learn More
Med Vet International Prascend, 1mg, 60 Tablets
Catalog No. 502828738
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Pergolide mesylate (1 mg tablets), a dopamine receptor agonist for oral use in horses. This is the first and only FDA-approved treatment for the control of clinical signs associated with Pituitary Pars Intermedia Dysfunction (Equine Cushings Disease) in horses. Do not use in horses intended for human consumption. May interfere with reproductive hormones and events in breeding, pregnant, or lactating horses.
Catalog No. 50-282-8738 Supplier Med Vet International Supplier No. RXPRASCEND160
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Pergolide mesylate (1 mg tablets), a dopamine receptor agonist for oral use in horses. This is the first and only FDA-approved treatment for the control of clinical signs associated with Pituitary Pars Intermedia Dysfunction (Equine Cushings Disease) in horses. Do not use in horses intended for human consumption. May interfere with reproductive hormones and events in breeding, pregnant, or lactating horses.