Learn More
Med Vet International Zoetis Anipryl Tablets, 30mg, 30ct.
Catalog No. 502828807
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Anipryl (selegiline hydrochloride, L-deprenyl hydrochloride) tablets are indicated for the control of clinical signs associated with canine cognitive dysfunction syndrome (CDS) and control of clinical signs associated with uncomplicated canine pituitary dependent hyperadrenocorticism (PDH).
Catalog No. 50-282-8807 Supplier Med Vet International Supplier No. RXANI3030
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Anipryl (selegiline hydrochloride, L-deprenyl hydrochloride) tablets are indicated for the control of clinical signs associated with canine cognitive dysfunction syndrome (CDS) and control of clinical signs associated with uncomplicated canine pituitary dependent hyperadrenocorticism (PDH).