Learn More
Med Vet International Zoetis Draxxin (Tulathromycin), 100mg, 50mL
Catalog No. 502828376
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Zoetis Draxxin is a ready-to-use sterile parenteral preparation containing tulathromycin, a semi-synthetic macrolide antibiotic of the subclass triamilide. For SQ injection in beef and non-lactating dairy cattle and IM injection in swine only.
Catalog No. 50-282-8376 Supplier Med Vet International Supplier No. RXDRAXXIN50ML
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Zoetis Draxxin is a ready-to-use sterile parenteral preparation containing tulathromycin, a semi-synthetic macrolide antibiotic of the subclass triamilide. For SQ injection in beef and non-lactating dairy cattle and IM injection in swine only.