Learn More
Med Vet International Zoetis Naxcel Sterile Powder, Injectable Antibiotic, 1gm
Catalog No. 502828461
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
NAXCEL (ceftiofur sodium) Sterile Powder is an Injectable antibiotic for treatment of bovine respiratory disease (BRD) and foot rot. Broad spectrum treatment: Treats the major causes of pneumonia and foot rot. Zero milk discard: Cows stay in the milking string. Short pre-slaughter withdrawal: When used according to label directions, you have the confidence of a four-day meat withholding time. Multiple-dose treatment options (3 to 5 days). Low-volume dose via subcutaneous (SC) or intramuscular (IM) administration
Catalog No. 50-282-8461 Supplier Med Vet International Supplier No. RXNAX1
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
NAXCEL (ceftiofur sodium) Sterile Powder is an Injectable antibiotic for treatment of bovine respiratory disease (BRD) and foot rot. Broad spectrum treatment: Treats the major causes of pneumonia and foot rot. Zero milk discard: Cows stay in the milking string. Short pre-slaughter withdrawal: When used according to label directions, you have the confidence of a four-day meat withholding time. Multiple-dose treatment options (3 to 5 days). Low-volume dose via subcutaneous (SC) or intramuscular (IM) administration