Learn More
Med Vet International Zoetis, Temaril-P 100 Tablets
Catalog No. 502828341
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Recommended for the relief of itching and inflammation commonly associated with most skin disorders of dogs such as the eczema. It has been found to be effective therapy and adjunctive therapy in various cough conditions of dogs such as kennel cough or tracheobronchitis, bronchitis including all allergic bronchitis and infections and coughs of nonspecific origin. Each tablet contains trimeprazine tartrate USP equivalent to trimeprazine, 5 mg, and prednisolone, 2 mg.
Catalog No. 50-282-8341 Supplier Med Vet International Supplier No. RXTEMARILP100
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Recommended for the relief of itching and inflammation commonly associated with most skin disorders of dogs such as the eczema. It has been found to be effective therapy and adjunctive therapy in various cough conditions of dogs such as kennel cough or tracheobronchitis, bronchitis including all allergic bronchitis and infections and coughs of nonspecific origin. Each tablet contains trimeprazine tartrate USP equivalent to trimeprazine, 5 mg, and prednisolone, 2 mg.