Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RY655 Mouse Anti-Human Ig, λ Light Chain, BD OptiBuild™

Catalog No. BDB773896 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB773896 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB773896 Supplier BD Biosciences Supplier No. 773896
Add to Cart
Add to Cart

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Human Samples. It is validated for Flow Cytometry.

The 1-155-2 monoclonal antibody specifically recognizes human immunoglobulin lambda light chains (Igλ). Immunoglobulins are found on B lymphocytes, plasma cells, and circulate in serum. The two light chains in an immunoglobulin are either kappa (Igκ) or Igλ. Immunoglobulins with Igλ are expressed on a minority of normal B lymphocytes and on neoplastic cells in some leukemias, lymphomas, and plasmacytomas. In serum, the 1-155-2 antibody binds to immunoglobulins with Igλ and free Igλ, but not to Igκ or immunoglobulin heavy chains.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone 1-155-2
Concentration 0.2 mg/mL
Conjugate RY655
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias IGL; IGL@; Ig λ; Immunoglobulin lambda
Host Species Mouse
Immunogen Human IgA1-λ myeloma Protein
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3535
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.