Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RY655 Mouse Anti-Human IgA1, BD OptiBuild™

Catalog No. BDB774062 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB774062 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB774062 Supplier BD Biosciences Supplier No. 774062
Add to Cart
Add to Cart

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Human Samples. It is validated for Flow Cytometry.

Monoclonal antibody that specifically recognizes the Fc region of human IgA1. Does not cross-react with other human IgH subclasses, including IgA2. Detects IgA1 expressed by plasmablasts, plasma cells, memory B cells, and some myeloma or plasmacytoma cells.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone B3506B4 (also known as DLDB4)
Concentration 0.2 mg/mL
Conjugate RY655
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias IGHA1; Immunoglobulin alpha 1
Host Species Mouse
Immunogen Human IgA1 Myeloma Protein
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3493
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.