Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RY655 Mouse Anti-Human IgG3, BD OptiBuild™

Catalog No. BDB774008 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB774008 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB774008 Supplier BD Biosciences Supplier No. 774008
Add to Cart
Add to Cart

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Human Samples. It is validated for Flow Cytometry.

The HP6050 monoclonal antibody specifically recognizes human Immunoglobulin G (IgG) subclass IgG3. It binds to the hinge region of the IgG3 heavy chain and does not crossreact with other immunoglobulin heavy chain subclasses. This antibody is used to detect soluble human IgG3, cytophilic IgG3 attached to Fc receptor-positive cells, or human IgG3 antibodies specifically bound to antigens.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone HP6050 (also known as CC4-DC10)
Concentration 0.2 mg/mL
Conjugate RY655
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias IGHG3; Ig gamma-3 chain C region; Immunoglobulin heavy constant gamma 3
Host Species Mouse
Immunogen Human IgG3 myeloma protein
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3502
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.