Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RY655 Mouse Anti-Mouse IgG1[a], BD OptiBuild™

Catalog No. BDB773492 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB773492 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB773492 Supplier BD Biosciences Supplier No. 773492
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Mouse Samples. It is validated for Flow Cytometry.

Specific for mouse IgG1 of Igh-C[a] and specific haplotypes. Dye: BD Horizon RealYellow 655 (RY655). Excitation maximum: 557 nm. Emission maximum: 654 nm. Designed to be excited by the Yellow-Green laser (561-nm). Suitable for both spectral and conventional cytometers. For research use only (RUO). Store undiluted at 4°C, protected from light. Do not freeze.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone 10.9
Concentration 0.2 mg/mL
Conjugate RY655
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Igh-4a; Ighg1; IgG1
Host Species Mouse
Immunogen Not reported
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 16017
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.