Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RY655 Mouse Anti-Syrian Hamster IgG2, BD OptiBuild™

Catalog No. BDB772327 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB772327 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB772327 Supplier BD Biosciences Supplier No. 772327
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Syrian Hamster Samples. It is validated for Flow Cytometry.

Reacts specifically with Syrian hamster IgG2 monoclonal antibodies, does not react with other hamster IgG groups or hamster IgM. Features BD Horizon RealYellow 655 (RY655) Dye, a tandem fluorochrome with excitation maximum at 557 nm and emission maximum at 654 nm. Usable on spectral and conventional cytometers, designed for excitation by Yellow-Green laser (561 nm). Can be used as an alternative to PE-Cy5 on conventional instruments with an optical filter centered near 660 nm. Usable with RB670 on conventional and spectral instruments equipped with Blue (488 nm) and Yellow-Green (561 nm) lasers.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Concentration 0.2 mg/mL
Conjugate RY655
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Pooled Armenian and Syrian Hamster IgG mAb
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Syrian Hamster
Content And Storage The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.